Learn More
TARGETMOL CHEMICALS INC GSK572A 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T274825MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GSK572A is a novel and potent potent inhibitor of EchA6 which can be used to treat tuberculosis. purity: 98%
Catalog No. 50-258-0607