Learn More
Selleck Chemical LLC GSK583
SDP
Catalog No. 5000002910
Encompass_Preferred
GSK583 is a highly potent and selective inhibitor of RIP2 kinase with IC50 of 5 nM GSK583 also inhibits both TNF- and IL-6 production with IC50 of 200 nM in explant cultures
Catalog No. 50-000-02910 Supplier Selleck Chemical LLC Supplier No. S82612MG
GSK583 is a highly potent and selective inhibitor of RIP2 kinase with IC50 of 5 nM GSK583 also inhibits both TNF- and IL-6 production with IC50 of 200 nM in explant cultures