Learn More
Selleck Chemical LLC GSK591 5mg 1616391-87-7 EPZ015866, GSK3203591
SDP
Supplier:  Selleck Chemical LLC S81115MG
Encompass_Preferred
GSK591 (EPZ015866, GSK3203591) is a potent selective inhibitor of the arginine methyltransferase PRMT5 with IC50 of 4 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1275695