Learn More
Medchemexpress LLC (-)-GSK598809 hydrochloride | 863766-31-8 | 517.97 | 5 MG
SDP
Catalog No. 5000402799
Encompass_Preferred
(-)-GSK598809 is an isomer of GSK598809, functioning as a potent and selective antagonist of the dopamine D3 Receptor (DRD3).
  • Potent and selective dopamine D3 Receptor (DRD3) antagonist.
  • Target: D3 Receptor.
  • Suitable for research applications.
  • Available in 5 mg quantity.
Catalog No. 50-004-02799 Supplier Medchemexpress LLC Supplier No. HY19654C5MG
(-)-GSK598809 is an isomer of GSK598809, functioning as a potent and selective antagonist of the dopamine D3 Receptor (DRD3).
  • Potent and selective dopamine D3 Receptor (DRD3) antagonist.
  • Target: D3 Receptor.
  • Suitable for research applications.
  • Available in 5 mg quantity.