Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (-)-GSK598809 hydrochloride | 863766-31-8 | 517.97 | 50 MG
SDP

Catalog No. 5000402800
Encompass_Preferred

(-)-GSK598809 is an isomer of GSK598809. GSK598809 is a potent and selective dopamine D3 Receptor (DRD3) antagonist.

  • Potent and selective dopamine D3 receptor (DRD3) antagonist
  • Target: D3 Receptor
  • Store at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month (sealed, away from moisture)

Catalog No. 50-004-02800 Supplier Medchemexpress LLC Supplier No. HY19654C50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(-)-GSK598809 is an isomer of GSK598809. GSK598809 is a potent and selective dopamine D3 Receptor (DRD3) antagonist.

  • Potent and selective dopamine D3 receptor (DRD3) antagonist
  • Target: D3 Receptor
  • Store at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month (sealed, away from moisture)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.