Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (-)-GSK598809 hydrochloride | 863766-31-8 | 99.6% | 10 MG
SDP

Catalog No. 5000294831
Encompass_Preferred

(-)-GSK598809 hydrochloride is the optically active isomer of GSK598809, a potent and selective dopamine D3 receptor (DRD3) antagonist supplied as a hydrochloride salt for research use. It is intended for pharmacological and receptor biology studies probing D3-mediated signaling.

  • Selective dopamine D3 receptor antagonist for receptor pharmacology
  • Hydrochloride salt form for improved solubility
  • High purity (99.6%) suitable for research applications
  • CAS number 863766-31-8 for substance identification
  • Molecular weight 517.97 g/mol
  • Chemical formula C22H24ClF4N5OS
  • Supplied as 10 MG research quantity

Catalog No. 50-002-94831 Supplier Medchemexpress LLC Supplier No. HY19654C10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(-)-GSK598809 hydrochloride is the optically active isomer of GSK598809, a potent and selective dopamine D3 receptor (DRD3) antagonist supplied as a hydrochloride salt for research use. It is intended for pharmacological and receptor biology studies probing D3-mediated signaling.

  • Selective dopamine D3 receptor antagonist for receptor pharmacology
  • Hydrochloride salt form for improved solubility
  • High purity (99.6%) suitable for research applications
  • CAS number 863766-31-8 for substance identification
  • Molecular weight 517.97 g/mol
  • Chemical formula C22H24ClF4N5OS
  • Supplied as 10 MG research quantity

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.