Learn More
Medchemexpress LLC (-)-GSK598809 hydrochloride | 863766-31-8 | 99.6% | 517.97 | 25 MG
SDP
Catalog No. 5000403160
Encompass_Preferred
(-)-GSK598809 is an isomer of GSK598809, which is a potent and selective dopamine D3 Receptor (DRD3) antagonist.
  • Potent and selective dopamine D3 receptor (DRD3) antagonist
  • Targets dopamine receptor
  • Involves GPCR/G protein and neuronal signaling pathways
  • Store at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-03160 Supplier Medchemexpress LLC Supplier No. 25MGHY19654C
(-)-GSK598809 is an isomer of GSK598809, which is a potent and selective dopamine D3 Receptor (DRD3) antagonist.
  • Potent and selective dopamine D3 receptor (DRD3) antagonist
  • Targets dopamine receptor
  • Involves GPCR/G protein and neuronal signaling pathways
  • Store at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month