Learn More
Selleck Chemical LLC GSK650394 5mg 890842-28-1
SDP
Supplier:  Selleck Chemical LLC S72095MG
Encompass_Preferred
GSK650394 is a serum- and glucocorticoid-regulated kinase-1 inhibitor with IC50 of 62 nM and 103 nM for SGK1 and SGK2, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0965343