Learn More
Medchemexpress LLC HY-107775 5mg Medchemexpress, GSK656 CAS:2131798-13-3 Purity:>98%
SDP
Catalog No. 501968465
Encompass_Preferred
Medchemexpress, HY-107775 5mg GSK656 CAS:2131798-13-3 GSK656 is a potent antitubercular agent, acting as an inhibitor of Mycobacterium tuberculosis (Mtb) leucyl-tRNA synthetase (LeuRS), with an IC50 of 0.2 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8465 Supplier Medchemexpress LLC Supplier No. HY1077755MG
Medchemexpress, HY-107775 5mg GSK656 CAS:2131798-13-3 GSK656 is a potent antitubercular agent, acting as an inhibitor of Mycobacterium tuberculosis (Mtb) leucyl-tRNA synthetase (LeuRS), with an IC50 of 0.2 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.