Learn More
Medchemexpress LLC HY-101192 100mg , GSK682753A CAS:1334294-76-6 Purity:98%
SDP
Catalog No. 501492862
Encompass_Preferred
Medchemexpress, HY-101192 100mg GSK682753A CAS:1334294-76-6 Formula:C23H21Cl3N2O3 IC50: 53.6 nM (EBI2) Purity:98% GSK682753A is a selective and highly potent inverse agonist of the epstein-barr virus-induced receptor 2 ( EBI2 ) with an IC 50 of 53.6 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2862 Supplier Medchemexpress LLC Supplier No. HY101192100MG
Medchemexpress, HY-101192 100mg GSK682753A CAS:1334294-76-6 Formula:C23H21Cl3N2O3 IC50: 53.6 nM (EBI2) Purity:98% GSK682753A is a selective and highly potent inverse agonist of the epstein-barr virus-induced receptor 2 ( EBI2 ) with an IC 50 of 53.6 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.