Learn More
AdipoGen GSK690693 (5 mg)
SDP
Supplier:  AdipoGen AGCR13628M005
Encompass_Preferred
Chemical. CAS: 937174-76-0. Formula: C21H27N7O3. MW: 425.5. pan-Akt inhibitor targeting Akt1, 2, 3 with IC50 values of 2, 13 and 9 nM, respectively. Inhibits AMPK, DAPK3, PAK4, 5 and 6, as well as members of the AGC kinase family including PKA, PrkX and PKC isoforms (IC50 < 100 nM). Occupies the ATP binding pocket of the kinase domain and competes with ATP. Anti-cancer compound both in vitro and in vivo. Blocks the phosphorylation of downstream targets of ATK in a dose-dependent manner, which leads to antiproliferative and apoptotic effects in tumor cell lines. Induces hyperglycemia related to peripheral insulin resistance, increased gluconeogenesis, and/or hepatic glycogenolysis.
Catalog No. 50-205-3998