Learn More
Medchemexpress LLC HY-136555 10mg , GSK717 CAS:1595278-21-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13655510MG
Encompass_Preferred
HY-136555 10mg GSK717 CAS: 1595278-21-9 GSK717 is a potent, selective NOD2 (nucleotide-binding oligomerization domain 2) inhibitor. GSK717 inhibits muramyl dipeptide (MDP)-induced NOD2-mediated signaling, with an IC50 of 400 nM for MDP-stimulated IL-8 secretion in HEK293/hNOD2 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0793