Learn More
Medchemexpress LLC HY-112152 5mg Medchemexpress, GSK726701A CAS:945721-87-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1121525MG
Encompass_Preferred
Medchemexpress, HY-112152 5mg GSK726701A CAS:945721-87-9 GSK726701A is a novel prostaglandin E2 receptor 4 (EP4) partial agonist with a pEC50 of 7.4. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0808