Learn More
Medchemexpress LLC HY-136570 5mg , GSK778 CAS:2451862-42-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1365705MG
Encompass_Preferred
HY-136570 5mg GSK778 CAS: 2451862-42-1 GSK778 (iBET-BD1) is a potent and selective BD1 bromodomain inhibitor of the BET proteins, with IC50s of 75 nM (BRD2 BD1), 41 nM (BRD3 BD1), 41 nM (BRD4 BD1), and 143 nM (BRDT BD1), respectively. GSK778 phenocopies the effects of pan-BET inhibitors in cancer models. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9099