Learn More
TARGETMOL CHEMICALS INC GSK778 HYDROCHLORIDE 5MG
SDP
Catalog No. 502578971
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GSK778 hydrochloride hydrochloride is a potent and selective BD1 bromodomain inhibitor of the BET proteins with IC50s of 75 nM (BRD2 BD1) 41 nM (BRD3 BD1) 41 nM (BRD4 BD1) and 143 nM (BRDT BD1) respectively. GSK778 hydrochloride hydrochloride phenocopies the effects of pan-BET inhibitors in cancer models[1]. purity: 98%
Catalog No. 50-257-8971 Supplier TARGETMOL CHEMICALS INC Supplier No. T9703L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GSK778 hydrochloride hydrochloride is a potent and selective BD1 bromodomain inhibitor of the BET proteins with IC50s of 75 nM (BRD2 BD1) 41 nM (BRD3 BD1) 41 nM (BRD4 BD1) and 143 nM (BRDT BD1) respectively. GSK778 hydrochloride hydrochloride phenocopies the effects of pan-BET inhibitors in cancer models[1]. purity: 98%