Learn More
Medchemexpress LLC HY-12776 5mg Medchemexpress, GSK805 CAS:1426802-50-7 Purity:>98%
SDP
Catalog No. 502030079
Encompass_Preferred
Medchemexpress, HY-12776 5mg GSK805 CAS:1426802-50-7 GSK805 is a potent, orally bioavailable, and CNS penetrant RORγt inhibitor with pIC50 of 8.4 and >8.2 for RORγ FRET assay and Th17 assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0079 Supplier Medchemexpress LLC Supplier No. HY127765MG
Medchemexpress, HY-12776 5mg GSK805 CAS:1426802-50-7 GSK805 is a potent, orally bioavailable, and CNS penetrant RORγt inhibitor with pIC50 of 8.4 and >8.2 for RORγ FRET assay and Th17 assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.