Learn More
Medchemexpress LLC HY-104021 10mg Medchemexpress, GSK840 CAS:2361146-30-5 Purity:>98%
SDP
Catalog No. 501968190
Encompass_Preferred
Medchemexpress, HY-104021 10mg GSK840 CAS:2361146-30-5 GSK840 (GSK'840) is a receptor-interacting protein kinase 3 (RIP3 or RIPK3) inhibitor, which binds RIP3 kinase domain with an IC50 of 0.9 nM, and inhibits kinase activity with an IC50 of 0.3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8190 Supplier Medchemexpress LLC Supplier No. HY10402110MG
Medchemexpress, HY-104021 10mg GSK840 CAS:2361146-30-5 GSK840 (GSK'840) is a receptor-interacting protein kinase 3 (RIP3 or RIPK3) inhibitor, which binds RIP3 kinase domain with an IC50 of 0.9 nM, and inhibits kinase activity with an IC50 of 0.3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.