Learn More
Medchemexpress LLC HY-107477 5mg Medchemexpress, GSK8573 CAS:1693766-04-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1074775MG
Encompass_Preferred
Medchemexpress, HY-107477 5mg GSK8573 CAS:1693766-04-9 GSK8573 (compound 23) is an inactive control compound for GSK2801. GSK8573 has binding activity to BRD9 with a Kd value of 1.04 μM and is inactive against BAZ2A/B and other bromodomain familiy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8421