Learn More
Medchemexpress LLC HY-111941 10mg Medchemexpress, GSK8612 CAS:2361659-62-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11194110MG
Encompass_Preferred
Medchemexpress, HY-111941 10mg GSK8612 CAS:2361659-62-1 GSK8612 is a highly selective and potent Tank-binding Kinase-1 (TBK1) inhibitor, with a pIC50 of 6.8 for recombinant TBK1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8989