Learn More
Medchemexpress LLC GSK8612 | 2361659-62-1 | 99.56% | 520.33 | 50 MG
SDP
Catalog No. 5000438976
Encompass_Preferred
GSK8612 is a highly selective and potent Tank-binding Kinase-1 (TBK1) inhibitor, with a pIC50 of 6.8 for recombinant TBK1. It is for research use only and not sold to patients.
  • Highly selective and potent Tank-binding Kinase-1 (TBK1) inhibitor.
  • Inhibits toll-like receptor (TLR)3-induced IRF3 phosphorylation in Ramos cells.
  • Inhibits type I IFN secretion in primary human mononuclear cells.
  • Inhibits secretion of IFNβ in response to dsDNA and cGAMP in THP1 cells.
Catalog No. 50-004-38976 Supplier Medchemexpress LLC Supplier No. HY11194150MG
GSK8612 is a highly selective and potent Tank-binding Kinase-1 (TBK1) inhibitor, with a pIC50 of 6.8 for recombinant TBK1. It is for research use only and not sold to patients.
  • Highly selective and potent Tank-binding Kinase-1 (TBK1) inhibitor.
  • Inhibits toll-like receptor (TLR)3-induced IRF3 phosphorylation in Ramos cells.
  • Inhibits type I IFN secretion in primary human mononuclear cells.
  • Inhibits secretion of IFNβ in response to dsDNA and cGAMP in THP1 cells.