Learn More
Medchemexpress LLC HY-111941 5mg Medchemexpress, GSK8612 CAS:2361659-62-1 Purity:>98%
SDP
Catalog No. 501932410
Encompass_Preferred
Medchemexpress, HY-111941 5mg GSK8612 CAS:2361659-62-1 GSK8612 is a highly selective and potent Tank-binding Kinase-1 (TBK1) inhibitor, with a pIC 50 of 6.8 for recombinant TBK1 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2410 Supplier Medchemexpress LLC Supplier No. HY1119415MG
Medchemexpress, HY-111941 5mg GSK8612 CAS:2361659-62-1 GSK8612 is a highly selective and potent Tank-binding Kinase-1 (TBK1) inhibitor, with a pIC 50 of 6.8 for recombinant TBK1 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.