Learn More
Medchemexpress LLC HY-19540 5mg Medchemexpress, GSK864 CAS:1816331-66-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY195405MG
Encompass_Preferred
Medchemexpress, HY-19540 5mg GSK864 CAS:1816331-66-4 GSK864 is an isocitrate dehydrogenase 1 (IDH1) mutant inhibitor; inhibits IDH1 mutants R132C, R132H, and R132G with IC50 values of 8.8, 15.2 and 16.6 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2141