Learn More
Medchemexpress LLC HY-100729 5mg Medchemexpress, GSK9311 CAS:1923851-49-3 Purity:>98%
SDP
Catalog No. 501931929
Encompass_Preferred
Medchemexpress, HY-100729 5mg GSK9311 CAS:1923851-49-3 GSK9311 is a potent inhibitor of the BRPF bromodomain with pIC 50 values of 6.0 and 4.3 for BRPF1 and BRPF2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1929 Supplier Medchemexpress LLC Supplier No. HY1007295MG
Medchemexpress, HY-100729 5mg GSK9311 CAS:1923851-49-3 GSK9311 is a potent inhibitor of the BRPF bromodomain with pIC 50 values of 6.0 and 4.3 for BRPF1 and BRPF2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.