Learn More
Medchemexpress LLC Gsk962 | 2049872-86-6 | 99.2% | 230.31 | C14H18N2O | 25 MG
SDP
Catalog No. 5000245945
Encompass_Preferred
GSK962 is the inactive enantiomer of GSK963 supplied as a purified small-molecule research reagent for use as a negative control to confirm on-target effects in studies of RIP kinase.
  • Inactive enantiomer control for RIP kinase studies.
  • High purity (99.18%) suitable for in vitro research.
  • Provided as a 25 MG powder for laboratory use.
  • Soluble in DMSO; follow SDS and datasheet for handling.
  • Certificate of analysis and SDS available for quality verification.
Catalog No. 50-002-45945 Supplier Medchemexpress LLC Supplier No. HY10302825MG
GSK962 is the inactive enantiomer of GSK963 supplied as a purified small-molecule research reagent for use as a negative control to confirm on-target effects in studies of RIP kinase.
  • Inactive enantiomer control for RIP kinase studies.
  • High purity (99.18%) suitable for in vitro research.
  • Provided as a 25 MG powder for laboratory use.
  • Soluble in DMSO; follow SDS and datasheet for handling.
  • Certificate of analysis and SDS available for quality verification.