Learn More
Medchemexpress LLC HY-103028 5mg Medchemexpress, GSK962 CAS:2049872-86-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1030285MG
Encompass_Preferred
Medchemexpress, HY-103028 5mg GSK962 CAS:2049872-86-6 GSK962 is an inactive enantiomer of GSK963 and can be used to confirm on-target effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8020