Learn More
Medchemexpress LLC HY-103028A 10mg Medchemexpress, GSK963 CAS:2049868-46-2 Purity:>98%
SDP
Catalog No. 501968021
Encompass_Preferred
Medchemexpress, HY-103028A 10mg GSK963 CAS:2049868-46-2 GSK963 is a chiral, highly potent and selective inhibitor of RIP1 kinase, with an IC50 of 29 nM. GSK963 is a selective and potent inhibitor of necroptosis in murine and human cells in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8021 Supplier Medchemexpress LLC Supplier No. HY103028A10MG
Medchemexpress, HY-103028A 10mg GSK963 CAS:2049868-46-2 GSK963 is a chiral, highly potent and selective inhibitor of RIP1 kinase, with an IC50 of 29 nM. GSK963 is a selective and potent inhibitor of necroptosis in murine and human cells in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.