Learn More
Medchemexpress LLC GSM III | 1493797-28-6 | 99.7% | 469.46 | 1 MG
SDP
Catalog No. 5000386183
Encompass_Preferred
GSM III is a γ-Secretase complexe (GSEC) modulator. It significantly regulates amyloid-β (Aβ) length at the extracellular interface between the protease (NCT, PSEN) and the substrate APPC99. GSM III can be used for Alzheimer's disease (AD) research.
  • Can be used for Alzheimer's disease (AD) research.
Catalog No. 50-003-86183 Supplier Medchemexpress LLC Supplier No. HY1714441MG
GSM III is a γ-Secretase complexe (GSEC) modulator. It significantly regulates amyloid-β (Aβ) length at the extracellular interface between the protease (NCT, PSEN) and the substrate APPC99. GSM III can be used for Alzheimer's disease (AD) research.
  • Can be used for Alzheimer's disease (AD) research.