Learn More
Carbosynth LLC. GsMTx-4 (0.1mg vial), 1ea, Disulfide bonds: (Disulfide bonds between Cys2-Cys17, Cys9-Cys23, and Cys16-Cys30), MW: 4095.8, Molecular formula: C185H273N49O45S6, CAS# 1209500-46-8
SDP
Supplier:  Carbosynth LLC. PCB4393S0.1MG
Encompass_Preferred
Synonyms: H-Gly-Cys-Leu-Glu-Phe-Trp-Trp-Lys-Cys-Asn-Pro-Asn-Asp-Asp-Lys-Cys-Cys-Arg-Pro-Lys-Leu-Lys-Cys-Ser-Lys-Leu-Phe-Lys-Leu-Cys-Asn-Phe-Ser-Phe-NH2, H-GCLEFWWKCNPNDDKCCRPKLKCSKLFKLCNFSF-NH2, Application: Inhibitor for Cation-Selective Stretch-Activated Channels /Atrial Fibrillation Inhibiting Peptide, Storage Temperature: -20°C, References: O.P. Hamill and B. Martinac, Physiol. Rev., 81, 685 (2001). (Review; MSCs), T.M. Suchyna, J.H. Johnson, K. Hamer, J.F. Leykam, D.A. Gage, H.F. Clemo, C.M. Baumgarten, and F. Sachs, J. Gen. Physiol., 115, 583 (2000). (Original), K.L. Ostrow, A. Mammoser, T. Suchyna, F. Sachs, R. Oswald, S. Kubo, N. Chino, and P. A. Gottlieb, Toxicon, 42, 263 (2003). (Primary structure; cDNA sequence & Pharmacol.)
Catalog No. 50-168-6262