Learn More
Medchemexpress LLC GsMTx4 | 1209500-46-8 | 99.9% | 4095.84 | 500 UG
SDP
Catalog No. 5000424387
Encompass_Preferred
GsMTx4 is a spider venom peptide that selectively inhibits cationic-permeable mechanosensitive channels (MSCs) belonging to the Piezo and TRP channel families. It also blocks cation-selective stretch-activated channels (SACs), and attenuates lysophosphatidylcholine (LPC)-induced astrocyte toxicity and microglial reactivity. GsMTx4 serves as a pharmacological tool for identifying the role of these excitatory MSCs in normal physiology and pathology.
  • Selectively inhibits cationic-permeable mechanosensitive channels
  • Blocks cation-selective stretch-activated channels
  • Attenuates lysophosphatidylcholine-induced astrocyte toxicity and microglial reactivity
  • Neuroprotective and inhibits demyelination
  • Reduces mechanical allodynia induced by inflammation and sciatic nerve injury
Catalog No. 50-004-24387 Supplier Medchemexpress LLC Supplier No. HYP1410C500UG
GsMTx4 is a spider venom peptide that selectively inhibits cationic-permeable mechanosensitive channels (MSCs) belonging to the Piezo and TRP channel families. It also blocks cation-selective stretch-activated channels (SACs), and attenuates lysophosphatidylcholine (LPC)-induced astrocyte toxicity and microglial reactivity. GsMTx4 serves as a pharmacological tool for identifying the role of these excitatory MSCs in normal physiology and pathology.
  • Selectively inhibits cationic-permeable mechanosensitive channels
  • Blocks cation-selective stretch-activated channels
  • Attenuates lysophosphatidylcholine-induced astrocyte toxicity and microglial reactivity
  • Neuroprotective and inhibits demyelination
  • Reduces mechanical allodynia induced by inflammation and sciatic nerve injury