Learn More
TARGETMOL CHEMICALS INC GSMTX4 TFA 1209500-46-8 5MG
SDP
Catalog No. 502578859
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg and bulk. Please contact Fisher for quotes. GsMTx4 TFA (1209500-46-8 free base) (GsMTx4 TFA) is a spider venom peptide that selectively inhibits cationic permeable mechanically-sensitive channels (MSCs) belonging to the Piezo and TRP channel families.
Catalog No. 50-257-8859 Supplier TARGETMOL CHEMICALS INC Supplier No. TP13005MG
Also available in 1 mg 2 mg 10 mg and bulk. Please contact Fisher for quotes. GsMTx4 TFA (1209500-46-8 free base) (GsMTx4 TFA) is a spider venom peptide that selectively inhibits cationic permeable mechanically-sensitive channels (MSCs) belonging to the Piezo and TRP channel families.