Learn More
Medchemexpress LLC GsMTx4 TFA | 99.9% | 4095.84 | 1 MG
SDP
Catalog No. 5000365889
Encompass_Preferred
GsMTx4 TFA is a spider venom peptide that selectively inhibits cationic-permeable mechanosensitive channels (MSCs) belonging to the Piezo and TRP channel families. It also blocks cation-selective stretch-activated channels (SACs) and attenuates lysophosphatidylcholine (LPC)-induced astrocyte toxicity and microglial reactivity, making it a pharmacological tool for identifying the role of these excitatory MSCs in normal physiology and pathology.
  • Selectively inhibits cationic-permeable mechanosensitive channels (MSCs)
  • Blocks cation-selective stretch-activated channels (SACs)
  • Attenuates lysophosphatidylcholine (LPC)-induced astrocyte toxicity
  • Attenuates microglial reactivity
  • Useful as a pharmacological tool for identifying the role of excitatory mechanosensitive channels
Catalog No. 50-003-65889 Supplier Medchemexpress LLC Supplier No. 1MGHYP1410A
GsMTx4 TFA is a spider venom peptide that selectively inhibits cationic-permeable mechanosensitive channels (MSCs) belonging to the Piezo and TRP channel families. It also blocks cation-selective stretch-activated channels (SACs) and attenuates lysophosphatidylcholine (LPC)-induced astrocyte toxicity and microglial reactivity, making it a pharmacological tool for identifying the role of these excitatory MSCs in normal physiology and pathology.
  • Selectively inhibits cationic-permeable mechanosensitive channels (MSCs)
  • Blocks cation-selective stretch-activated channels (SACs)
  • Attenuates lysophosphatidylcholine (LPC)-induced astrocyte toxicity
  • Attenuates microglial reactivity
  • Useful as a pharmacological tool for identifying the role of excitatory mechanosensitive channels