Learn More
Medchemexpress LLC GsMTx4 TFA | 99.9% | 4095.84 (free base) | 5 MG
SDP
Catalog No. 5000423185
Encompass_Preferred
GsMTx4 TFA is a spider venom peptide designed to selectively inhibit cationic-permeable mechanosensitive channels (MSCs) within the Piezo and TRP channel families. It acts as a pharmacological tool to investigate the role of these excitatory MSCs in both normal physiological processes and various pathological conditions.
  • Selectively inhibits cationic-permeable mechanosensitive channels (MSCs) belonging to Piezo and TRP families.
  • Blocks cation-selective stretch-activated channels in various cell types.
  • Attenuates lysophosphatidylcholine-induced astrocyte toxicity and microglial reactivity.
  • Neuroprotective in the cerebral cortex.
  • Reduces mechanical allodynia in models of inflammation and sciatic nerve injury.
Catalog No. 50-004-23185 Supplier Medchemexpress LLC Supplier No. HYP1410A5MG
GsMTx4 TFA is a spider venom peptide designed to selectively inhibit cationic-permeable mechanosensitive channels (MSCs) within the Piezo and TRP channel families. It acts as a pharmacological tool to investigate the role of these excitatory MSCs in both normal physiological processes and various pathological conditions.
  • Selectively inhibits cationic-permeable mechanosensitive channels (MSCs) belonging to Piezo and TRP families.
  • Blocks cation-selective stretch-activated channels in various cell types.
  • Attenuates lysophosphatidylcholine-induced astrocyte toxicity and microglial reactivity.
  • Neuroprotective in the cerebral cortex.
  • Reduces mechanical allodynia in models of inflammation and sciatic nerve injury.