Learn More
Biomatik Corporation Gsn Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11886
Encompass_Preferred
Gsn Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to GSN. The Gsn Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Gelsolin Protein (28-782Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: GSN. Accession Number: P06396. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6732