Learn More
Enzo Life Sciences GSNO (100mg). CAS: 57564-91-7
SDP
Supplier:  Enzo Life Sciences ALX420002M100
Encompass_Preferred
NO donor which spontaneously releases NO under physiological conditions. Half-life of 159 hours under physiological conditions. Selectively inhibits platelet activation and displays cardioprotective effects. Inhibits NF-κB activation. Inhibits endothelial cell proliferation and induces apoptosis in T cells. Alternative name: S-Nitroso-L-glutathione, N-(N-L-gamma-Glutamyl-S-nitroso-L-cysteinyl)-glycine. Purity: ≥98% (HPLC). Appearance: Crystalline solid. Solubility: Soluble in water (25mg/ml),  DMSO (20mg/ml) or PBS; sparingly soluble in 100% ethanol, methanol or dimethyl formamide. Long Term Storage: -20°C.
Catalog No. 50-200-9094