Learn More
Medchemexpress LLC GSPT1 protein, human (biotinylated, Avi tag) | 92.0% | 10UG
SDP
Catalog No. 5000175748
Encompass_Preferred
Recombinant human GSPT1 protein is expressed in Sf9 insect cells and site-specifically biotinylated via an Avi tag. Supplied as a 0.22 μm filtered solution in 50 mM HEPES, pH 7.5, with 1 mM DTT, 200 mM NaCl, and 20% glycerol, the product is provided frozen and shipped on dry ice for storage at -80°C.
  • Human-derived sequence spanning residues D2-D637; approx. 70.8 kDa.
  • Site-specific Avi biotinylation for consistent streptavidin-based applications.
  • High purity (92.0% by reducing SDS-PAGE) suitable for biochemical assays.
  • Formulated with glycerol and reducing agent for enhanced stability.
  • Available in small-scale sizes to support assay development and screening.
  • Shipped on dry ice; recommended long-term storage at -80°C.
Catalog No. 50-001-75748 Supplier Medchemexpress LLC Supplier No. HYP70314110UG
Recombinant human GSPT1 protein is expressed in Sf9 insect cells and site-specifically biotinylated via an Avi tag. Supplied as a 0.22 μm filtered solution in 50 mM HEPES, pH 7.5, with 1 mM DTT, 200 mM NaCl, and 20% glycerol, the product is provided frozen and shipped on dry ice for storage at -80°C.
  • Human-derived sequence spanning residues D2-D637; approx. 70.8 kDa.
  • Site-specific Avi biotinylation for consistent streptavidin-based applications.
  • High purity (92.0% by reducing SDS-PAGE) suitable for biochemical assays.
  • Formulated with glycerol and reducing agent for enhanced stability.
  • Available in small-scale sizes to support assay development and screening.
  • Shipped on dry ice; recommended long-term storage at -80°C.