Learn More
Aviva Systems Biology Corp GSPT2 Antibody - N-terminal region (ARP55356_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55356_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GSPT2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GSPT2. Alternate names: FLJ10441, GST2, eRF3b, ERF3BApplicationWB. Species reactivity: Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-61478