Learn More
Aviva Systems Biology Corp GSR Antibody - N-terminal region (ARP54344_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54344_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GSR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GSR. Alternate names: MGC78522ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61180