Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec GSR Collection Kit, 2 Stubs
SDP

Catalog No. 22446270 Shop All Mopec Products
Change view
Click to view available options
Quantity:
10/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-446-270 10/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-446-270 Supplier Mopec Supplier No. FS480
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to collect gunshot residue from a subject's hands or nearby surfaces, these sampling materials support laboratory analysis and secure evidence handling. Depending on the configuration, components support AA or SEM collection, sampling, tamper evidence, preservation, and chain of custody.

Construction

  • (1) Tamper-evident, pre-sealed Kit Envelope
  • Kit instruction sheet
  • (1) Gunshot residue analysis information form
  • (1) Pair of disposable plastic gloves
  • (1) Right hand vial with cap containing an SEM stub covered with double-sided adhesive carbon tape
  • (1) Left hand vial with cap containing an SEM stub covered with double-sided adhesive carbon tape
  • (1) Tamper-evident police evidence seal for resealing kit after evidence collection

Dimensions

  • Envelope: 6 L x 9 W in.

Quantity

  • 10 per case

Performance

  • For agencies whose crime labs perform GSR analysis by SEM on two collection stubs

Specifications

Dimensions (L x W) 6 x 9 in.
Product Type GSR Collection Kit
Quantity 10/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.