Learn More
Mopec GSR Collection Kit, 4 Stubs
SDP
Catalog No. 22446271 Shop All Mopec Products
Change view
Click to view available options
Quantity:
10/Cs.
Catalog No. Quantity
22-446-271 10/Cs.
1 options
Catalog No. 22-446-271 Supplier Mopec Supplier No. FS482
Add to Cart
Add to Cart
Designed to collect gunshot residue from a subject's hands or nearby surfaces, these sampling materials support laboratory analysis and secure evidence handling. Depending on the configuration, components support AA or SEM collection, sampling, tamper evidence, preservation, and chain of custody.
Construction
  • (1) Tamper-evident, pre-sealed kit envelope
  • Kit instruction sheet
  • (1) Gunshot residue analysis information form
  • (1) Pair of disposable plastic gloves
  • (1) Right and left back vials with cap containing a SEM stub covered with double-sided adhesive carbon tape
  • (1) Right and left palm vials with cap containing a SEM stub covered with double-sided adhesive carbon tape
  • (1) Tamper-evident police evidence seal for resealing kit after evidence collection
Dimensions
  • Envelope: 6 L x 9 W in.
Quantity
  • 10 per case
Performance
  • For law enforcement agencies whose crime labs use SEM as their method of GSR analysis request 'Right Back', 'Left Back', 'Right Palm', and 'Left Palm' samples be collected from the suspect’s hands
Specifications
Product Type GSR Collection Kit
Quantity 10/Cs.