Learn More
MSC GSS Safety Vest, ANSI Class 2, 5X-Large, Orange, 1 ea
Catalog No. 5000792690
Encompass
The GSS Safety Premium Class 2 Hyper Lite Vest is a high-visibility safety vest constructed from 100% polyester smooth mesh, featuring a zipper closure. Designed for general purpose use, this vest meets ANSI/ISEA 107-2015 Class 2 certification for enhanced worker safety.
  • Zipper closure
  • Made from 100% polyester smooth mesh material
  • Features 2 in reflective tape with brilliant trim
  • Reflective edging for extra visibility
  • Two lower outside cargo pockets with grommets and flaps
  • Two inside iPad pockets
  • Reinforced webbing
  • Equipped with 6 total pockets
Catalog No. 50-007-92690 Supplier MSC Supplier No. 82662875
The GSS Safety Premium Class 2 Hyper Lite Vest is a high-visibility safety vest constructed from 100% polyester smooth mesh, featuring a zipper closure. Designed for general purpose use, this vest meets ANSI/ISEA 107-2015 Class 2 certification for enhanced worker safety.
  • Zipper closure
  • Made from 100% polyester smooth mesh material
  • Features 2 in reflective tape with brilliant trim
  • Reflective edging for extra visibility
  • Two lower outside cargo pockets with grommets and flaps
  • Two inside iPad pockets
  • Reinforced webbing
  • Equipped with 6 total pockets