Learn More
Genscript Corporation GST Fusion Protein Purification Kit
Catalog No. NC1348277
Encompass
Glutathione Resin (Cat. No. L00206) is an affinity chromatography medium designed for easy, one-step purification of recombinant glutathione S-transferase (GST) fusion proteins and other glutathione binding proteins expressed in E. coli, insect cells and mammalian cells. The recombinant GST fusion proteins can be purified directly from pre-treated cell lysate using Glutathione Resin. It is the excellent choice for high performance purifications.
Catalog No. NC1348277 Supplier Genscript Corporation Supplier No. L00207
Glutathione Resin (Cat. No. L00206) is an affinity chromatography medium designed for easy, one-step purification of recombinant glutathione S-transferase (GST) fusion proteins and other glutathione binding proteins expressed in E. coli, insect cells and mammalian cells. The recombinant GST fusion proteins can be purified directly from pre-treated cell lysate using Glutathione Resin. It is the excellent choice for high performance purifications.