Learn More
Cayman Chemical GST GlutathIon 1ea
SDP
Catalog No. 502893888
Encompass_Preferred
An NQO1 substrate and a derivative ofBlapachone converted by NQO1 to NAD and increases the ratio of NAD to NADH in a cell-free assay as well as C2C12 and L6 myoblasts at 1 1 or 2 UM respectively increases intracellular ATP levels as well as decreases intracellular lactate and ROS levels in primary fibroblasts isolated from patients with MELAS at 1 UM increases the levels of mitochondrial complex I and mitochondrial complex III as well as increases the OCR in primary fibroblasts isolated from patients with MELAS at 1 UM inhibits cisplatin-induced apoptosis production of mitochondrial ROS and mitochondrial membrane depolarization in primary mouse cochlear organ of Corti cells at 1 UM
Catalog No. 50-289-3888 Supplier Cayman Chemical Supplier No. 7033161 EA
An NQO1 substrate and a derivative ofBlapachone converted by NQO1 to NAD and increases the ratio of NAD to NADH in a cell-free assay as well as C2C12 and L6 myoblasts at 1 1 or 2 UM respectively increases intracellular ATP levels as well as decreases intracellular lactate and ROS levels in primary fibroblasts isolated from patients with MELAS at 1 UM increases the levels of mitochondrial complex I and mitochondrial complex III as well as increases the OCR in primary fibroblasts isolated from patients with MELAS at 1 UM inhibits cisplatin-induced apoptosis production of mitochondrial ROS and mitochondrial membrane depolarization in primary mouse cochlear organ of Corti cells at 1 UM