Learn More
Enzo Life Sciences Glutathione S-Transferase monoclonal antibody (8-326) (biotin conjugate) (50µl)
SDP
Supplier:  Enzo Life Sciences ENZABS2800050
Encompass_Preferred
Clone: 8-326. Isotype: IgG. Immunogen: Full-length recombinant human Glutathione S-transferase. Species reactivity: Species independent. Applications: IP, WB. Purity: Purified. Formulation: Liquid. In PBS containing a proprietary stabilizer and 0.02% sodium azide.
Catalog No. 50-201-2823