Learn More
Enzo Life Sciences Glutathione S-Transferase monoclonal antibody (8C1-G4) (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905618100
Encompass_Preferred
Clone: 8C1-G4. Host: Mouse. Isotype: IgG2b. Immunogen: GST fusion protein. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Species independent. Applications: ELISA, IP, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 0.01% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-6967