Learn More
Enzo Life Sciences GST P polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences ADIMSA101E
Encompass_Preferred
Host: Rabbit. Immunogen: Native rat liver GST-P. UniProt: P04906. Species Reactivity: Human, Mouse, Rat. Applications: IHC, WB. Recommended dilutions: Immunohistochemistry (1:1,000-1:2,000). Western Blot (1:1,000-1:2,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol.
Catalog No. 50-666-973