Learn More
AdipoGen GST pAb IG-727
SDP
Supplier:  AdipoGen AG25T0115PFC200
Encompass_Preferred
Polyclonal Antibody. Recognizes GST. Source Rabbit. Applications WB. Liquid. Sterile filtered. Best known for their ability to conjugate glutathione GSH and thereby detoxify cellular environments, GSTs are also capable of binding nonsubstrate ligands with important cell signaling implications. GST is used as a purification tag for recombinant proteins.
Catalog No. 50-256-3159