Learn More
Enzo Life Sciences GST-pi polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences ADIMSA102E
Encompass_Preferred
Host: Rabbit. Immunogen: Human placental GST-π. UniProt: P09211. Species Reactivity: Human, Mouse, Rat. Applications: IHC, WB. Recommended dilutions: Immunohistochemistry (1:500-1:1,000). Western Blot (1:500-1:1,000). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol.
Catalog No. 50-666-974