Learn More
Enzo Life Sciences Glutathione S-transferase-Pi monoclonal antibody (USal-hGST-Pi-McAb-1) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804510C100
Encompass_Preferred
Clone: USal-hGST-Pi-McAb-1. Isotype: IgG1. Immunogen: Recombinant human placenta GST-Pi (glutathione S-transferase-Pi). Species reactivity: Human, Rat Leishmania. Applications: IHC (FS), WB. Purity: Protein G-affinity purified. Formulation: Liquid. Contains 100nM sodium azide and 0.7% BSA.
Catalog No. 50-200-9934