Learn More
Sino Biological Recombinant Schistosoma japonicum GST Protein
Supplier:  Sino Biological 11213HNAE1
Encompass_Preferred
A DNA sequence encoding the Glutathione S-transferase (P08515-1) (Met1-Lys218) was expressed.
Catalog No. 50-255-4790