Learn More
Thermo Scientific™ GST-Tag Magnetic Agarose Purification Bundle
Description
Includes
Pierce Glutathione Magnetic Agarose Beads (1 mL), Halt Protease Inhibitor Cocktail (100X) (1 mL), B-PER Complete Bacterial Protein Extraction Reagent (250 mL)
- Thermo Scientific Pierce Glutathione Magnetic Agarose Beads provide a fast, convenient method for purification of glutathione-S-transferase (GST)-tagged fusion proteins from a bacterial, yeast, or mammalian crude cell lysate. Pierce Glutathione Magnetic Agarose Beads are well suited for purifying GST fusion proteins from a soluble protein extract and are ideal for the purification of proteins expressed at low levels from diluted supernatants
- Thermo Scientific B-PER Complete Bacterial Protein Extraction Reagent is an easy-to-use cell lysis reagent. It is a nonionic detergent-based solution that effectively disrupts cells and solubilizes native or recombinant proteins without denaturation. B-PER Complete Bacterial Protein Extraction Reagent is an all-inclusive formulation that combines the lysis reagent with lysozyme and a universal nuclease to enable mild extraction from both gram-negative and gram-positive bacteria
- Thermo Scientific Halt Protease Inhibitor Cocktail combined with this lysis buffer formulation added before use, protects proteins from proteolytic degradation through inhibition of serine proteases, cysteine proteases, aspartic acid proteases, and aminopeptidases that are typically present in cell lysate samples
Specifications
Specifications
| Quantity | 1 Bundle |
| Includes | Pierce Glutathione Magnetic Agarose Beads (1 mL), Halt Protease Inhibitor Cocktail (100X) (1 mL), B-PER Complete Bacterial Protein Extraction Reagent (250 mL) |
| Content And Storage | See instructions for individual components |
| Matrix | Magnetic Agarose |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.